Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDF1
Entrez Gene ID
417286
Gene Name
endothelial differentiation related factor 1
Gene Aliases
-
Chromosome Location
Chr.17: 913889 - 917066 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006203.1 NP_001006203.1 2 - 3 158 117
GenBank mRNA AJ719464.1 - 2 - 3 158 117

View Full Product Details