Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YWHAE
Entrez Gene ID
417554
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein epsilon
Gene Aliases
-
Chromosome Location
Chr.19: 5331007 - 5350911 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006219.1 NP_001006219.1 5 - 6 827 66
GenBank mRNA AJ719304.1 - 5 - 6 827 66

View Full Product Details