Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WASH1
Entrez Gene ID
418145
Gene Name
WAS protein family homolog 1
Gene Aliases
FAM39B, WASH2P, WASHC1
Chromosome Location
Chr.1: 59938866 - 59978105 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006245.1 NP_001006245.1 5 - 6 565 64
XM_015285752.1 XP_015141238.1 6 - 7 897 64
XM_025148817.1 XP_025004585.1 5 - 6 618 64
XM_025148818.1 XP_025004586.1 5 - 6 579 64
XM_025148819.1 XP_025004587.1 7 - 8 1038 64
GenBank mRNA AJ720178.1 - 5 - 6 565 64

View Full Product Details