Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WASH1
Entrez Gene ID
418145
Gene Name
WAS protein family homolog 1
Gene Aliases
FAM39B, WASH2P, WASHC1
Chromosome Location
Chr.1: 59938866 - 59978105 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006245.1 NP_001006245.1 9 - 10 1244 84
XM_015285752.1 XP_015141238.1 10 - 11 1576 84
XM_025148817.1 XP_025004585.1 9 - 10 1297 84
XM_025148818.1 XP_025004586.1 9 - 10 1258 84
XM_025148819.1 XP_025004587.1 11 - 12 1717 84
GenBank mRNA AJ720178.1 - 9 - 10 1244 84

View Full Product Details