Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FKBP4
Entrez Gene ID
418261
Gene Name
FK506 binding protein 4
Gene Aliases
-
Chromosome Location
Chr.1: 75399374 - 75413494 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006250.1 NP_001006250.1 6 - 7 899 101
GenBank mRNA AJ720350.1 - 6 - 7 899 101

View Full Product Details