Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC18
Entrez Gene ID
419575
Gene Name
zinc finger, DHHC-type containing 18
Gene Aliases
-
Chromosome Location
Chr.23: 1890574 - 1902901 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006310.2 NP_001006310.2 6 - 7 664 96
XM_015297547.2 XP_015153033.2 6 - 7 1918 96
XM_015297549.2 XP_015153035.2 6 - 7 898 96
XM_025142984.1 XP_024998752.1 6 - 7 902 96
GenBank mRNA AJ720991.1 - 6 - 7 664 95

View Full Product Details