Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASNSD1
Entrez Gene ID
423983
Gene Name
asparagine synthetase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 352533 - 359220 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006509.1 NP_001006509.1 5 - 6 1978 130
GenBank mRNA AJ720404.1 - 5 - 6 1978 130

View Full Product Details