Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC13
Entrez Gene ID
427165
Gene Name
trafficking protein particle complex 13
Gene Aliases
C5orf44, CZH5ORF44, Trs65-related
Chromosome Location
Chr.Z: 20606667 - 20631170 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006577.2 NP_001006577.2 4 - 5 357 148
NM_001194996.1 NP_001181925.1 4 - 5 357 148
XM_015277311.2 XP_015132797.1 4 - 5 374 148
XM_015277312.2 XP_015132798.1 4 - 5 388 148
GenBank mRNA AJ720892.1 - 4 - 5 358 148
BU335726.1 - 4 - 5 330 148
DT656730.1 - 2 - 3 124 148

View Full Product Details