Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPL4
Entrez Gene ID
415551
Gene Name
ribosomal protein L4
Gene Aliases
-
Chromosome Location
Chr.10: 18943500 - 18948821 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007479.1 NP_001007480.1 3 - 4 341 73
GenBank mRNA AJ720803.1 - 3 - 4 341 73
AY389964.1 - 2 - 3 229 73

View Full Product Details