Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIAA1024
Entrez Gene ID
415485
Gene Name
KIAA1024
Gene Aliases
DD1
Chromosome Location
Chr.10: 13188588 - 13207553 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007827.2 NP_001007828.1 1 - 2 2352 63
XM_015292000.2 XP_015147486.1 2 - 3 3337 63
XM_015292001.2 XP_015147487.1 3 - 4 3482 63
GenBank mRNA AY712943.1 - 1 - 2 2356 63

View Full Product Details