Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TANGO2
Entrez Gene ID
416784
Gene Name
transport and golgi organization 2 homolog
Gene Aliases
C15H22ORF25, C22orf25
Chromosome Location
Chr.15: 1557612 - 1575120 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007836.1 NP_001007837.1 4 - 5 385 78
XM_015275249.2 XP_015130735.1 4 - 5 385 78
XM_015275250.2 XP_015130736.1 4 - 5 461 78
XM_015275251.2 XP_015130737.1 4 - 5 512 78
XM_015275252.2 XP_015130738.1 4 - 5 593 78
XM_015275253.2 XP_015130739.1 4 - 5 525 78
XM_025155442.1 XP_025011210.1 4 - 5 1517 78
GenBank mRNA AJ720940.1 - 4 - 5 385 78

View Full Product Details