Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSU72
Entrez Gene ID
419410
Gene Name
SSU72 homolog, RNA polymerase II CTD phosphatase
Gene Aliases
-
Chromosome Location
Chr.21: 2122367 - 2144383 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007875.1 NP_001007876.1 3 - 4 439 85
GenBank mRNA AJ720377.1 - 3 - 4 439 85

View Full Product Details