Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A36
Entrez Gene ID
424817
Gene Name
solute carrier family 25 member 36
Gene Aliases
-
Chromosome Location
Chr.9: 6918245 - 6948708 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007960.1 NP_001007961.1 3 - 4 381 77
GenBank mRNA AJ720037.1 - 3 - 4 381 77

View Full Product Details