Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASSF5
Entrez Gene ID
419844
Gene Name
Ras association domain family member 5
Gene Aliases
-
Chromosome Location
Chr.26: 2489020 - 2506554 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008452.1 NP_001008452.1 - 672 79
XM_015298925.2 XP_015154411.1 - 932 79
XM_015298926.2 XP_015154412.1 - 814 79
XM_015298927.2 XP_015154413.1 - 614 79
GenBank mRNA AJ720993.1 - - 672 79

View Full Product Details