Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1H21ORF91
Entrez Gene ID
395489
Gene Name
chromosome 1 open reading frame, human C21orf91
Gene Aliases
C21orf91
Chromosome Location
Chr.1: 100750280 - 100767071 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204737.1 NP_990068.1 4 - 5 773 129
XM_015295756.2 XP_015151242.1 4 - 5 753 129
XM_015295763.2 XP_015151249.1 4 - 5 804 129
GenBank mRNA AF162861.1 - 4 - 5 773 129

View Full Product Details