Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOUL
Entrez Gene ID
395567
Gene Name
SOUL protein
Gene Aliases
-
Chromosome Location
Chr.5: 58929202 - 58939828 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204789.1 NP_990120.1 3 - 4 464 89
GenBank mRNA AF117612.1 - 3 - 4 464 89

View Full Product Details