Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNNC2
Entrez Gene ID
396434
Gene Name
troponin C2, fast skeletal type
Gene Aliases
-
Chromosome Location
Chr.20: 10510924 - 10512722 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205450.2 NP_990781.1 5 - 6 496 92
GenBank mRNA M19027.1 - 5 - 6 500 92

View Full Product Details