Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPL6
Entrez Gene ID
373957
Gene Name
ribosomal protein L6
Gene Aliases
-
Chromosome Location
Chr.15: 6679237 - 6682601 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204152.1 NP_989483.1 5 - 6 584 140
GenBank mRNA AY032864.1 - 5 - 6 584 140

View Full Product Details