Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
SIX1
Entrez Gene ID
693262
Gene Name
SIX homeobox 1
Gene Aliases
-
Chromosome Location
Chr.5: 54593128 - 54595592 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044685.1 NP_001038150.1 2 - 2 752 74
GenBank mRNA AB199734.1 - 2 - 2 752 74

View Full Product Details