Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXG1
Entrez Gene ID
396110
Gene Name
forkhead box G1
Gene Aliases
CBF-1
Chromosome Location
Chr.5: 33732830 - 33741745 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205193.1 NP_990524.1 1 - 1 1556 100
XM_025150606.1 XP_025006374.1 3 - 3 6430 100
GenBank mRNA L36814.1 - 1 - 1 1310 100
U47275.1 - 1 - 1 1556 100

View Full Product Details