Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GH1
Entrez Gene ID
547502
Gene Name
GH1 protein
Gene Aliases
-
Chromosome Location
Chr.1: 1886333 - 1890391 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF016633.1 - 2 - 3 544 66

View Full Product Details