Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
URE
Entrez Gene ID
547672
Gene Name
urease
Gene Aliases
-
Chromosome Location
Chr.11: 38418308 - 38425109 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249285.1 NP_001236214.1 14 - 15 1917 81
GenBank mRNA AJ276866.1 - 15 - 16 2091 81
AY230156.1 - 14 - 15 1917 81

View Full Product Details