Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDH
Entrez Gene ID
547749
Gene Name
proline dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.19: 6088361 - 6091617 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250359.1 NP_001237288.1 2 - 3 1215 81
GenBank mRNA AY492003.1 - 2 - 3 1215 81

View Full Product Details