Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNAP-1
Entrez Gene ID
547812
Gene Name
nucleosome assembly protein 1
Gene Aliases
-
Chromosome Location
Chr.15: 15484078 - 15487474 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251385.1 NP_001238314.1 3 - 4 264 70
GenBank mRNA BT096713.1 - 3 - 4 297 70
BT098302.1 - 3 - 4 295 70
L38856.1 - 3 - 4 264 70

View Full Product Details