Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC48
Entrez Gene ID
547850
Gene Name
plamsma membrane-associated AAA-ATPase
Gene Aliases
VCP
Chromosome Location
Chr.13: 40364413 - 40369915 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001248170.1 NP_001235099.1 7 - 8 1819 78
GenBank mRNA AK286854.1 - - 3164 78
DQ359725.1 - 7 - 8 1819 78
U20213.1 - 7 - 8 1839 78

View Full Product Details