Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFR2
Entrez Gene ID
732645
Gene Name
beta-glycosidase-like
Gene Aliases
-
Chromosome Location
Chr.17: 2710451 - 2715424 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251399.1 NP_001238328.1 3 - 4 643 87
GenBank mRNA AM238658.1 - 3 - 4 643 87

View Full Product Details