Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FUT13
Entrez Gene ID
548046
Gene Name
protein-O-fucosyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.16: 1291130 - 1294325 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ831835.1 - 2 - 3 318 90

View Full Product Details