Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WRKY24
Entrez Gene ID
100127377
Gene Name
WRKY24 protein
Gene Aliases
-
Chromosome Location
Chr.17: 6042706 - 6050500 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK245832.1 - 4 - 5 1341 67
EU019566.1 - 2 - 3 649 67

View Full Product Details