Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY25
Entrez Gene ID
100127378
Gene Name
WRKY25 protein
Gene Aliases
-
Chromosome Location
Chr.8: 898825 - 900025 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250081.1 NP_001237010.1 1 - 2 383 93
GenBank mRNA EU019567.1 - 1 - 2 383 93

View Full Product Details