Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALAAT1
Entrez Gene ID
100127413
Gene Name
alanine aminotransferase 1
Gene Aliases
-
Chromosome Location
Chr.16: 1199953 - 1205724 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250624.1 NP_001237553.1 7 - 8 588 97
GenBank mRNA EU165371.1 - 7 - 8 588 97

View Full Product Details