The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251781.2 | NP_001238710.1 | 3 - 4 | 909 | 62 |
GenBank mRNA | AB359233.1 | - | 3 - 4 | 960 | 62 |