Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WNK6
Entrez Gene ID
100272197
Gene Name
with no lysine kinase 6
Gene Aliases
-
Chromosome Location
Chr.7: 4631644 - 4635415 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249016.1 NP_001235945.1 4 - 5 492 93
GenBank mRNA FJ795036.1 - 4 - 5 492 93

View Full Product Details