The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001249091.1 | NP_001236020.1 | 1 - 2 | 52 | 94 |
GenBank mRNA | FJ795038.1 | - | 1 - 2 | 52 | 94 |