Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WNK8
Entrez Gene ID
100272202
Gene Name
with no lysine kinase 8
Gene Aliases
-
Chromosome Location
Chr.20: 22785968 - 22788810 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249091.1 NP_001236020.1 2 - 3 91 82
GenBank mRNA FJ795038.1 - 2 - 3 91 82

View Full Product Details