The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001250337.1 | NP_001237266.1 | 1 - 1 | 348 | 104 |
GenBank mRNA | DQ787068.1 | - | 1 - 1 | 348 | 104 |