Gene Transcripts
- Gene Symbol
- BMP5
- Entrez Gene ID
- 653
- Gene Name
- bone morphogenetic protein 5
- Gene Aliases
- Chromosome Location
- Chr. 6: 55753653 - 55875625 on Build GRCh38
- UniGene ID
- Hs.296648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021073.2 |
NP_066551.1
|
3, 4 |
1549 |
|
XM_005249304.3 |
XP_005249361.1
|
3, 4 |
1597 |
|
XM_011514816.2 |
XP_011513118.1
|
3, 4 |
845 |
|
XM_011514817.2 |
XP_011513119.1
|
3, 4 |
1597 |
|
XM_017011198.1 |
XP_016866687.1
|
2, 3 |
254 |
|
GenBank mRNA |
AK291559.1 |
- |
3, 4 |
1562 |
|
AK303576.1 |
- |
3, 4 |
851 |
|
BC027958.1 |
- |
3, 4 |
1549 |
5209637 |
KC485577.1 |
- |
3, 4 |
837 |
|
M60314.1 |
- |
3, 4 |
1535 |
|
View Full Product Details