Gene Transcripts

Gene Symbol
MYRF
Entrez Gene ID
745
Gene Name
myelin regulatory factor
Gene Aliases
C11orf9, MRF, Ndt80, pqn-47
Chromosome Location
Chr. 11: 61752617 - 61788518 on Build GRCh38
UniGene ID
Hs.473109
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127392.2 NP_001120864.1 27 3540
NM_013279.3 NP_037411.1 26 3347
XM_005274222.1 XP_005274279.1 27 3531
XM_005274223.1 XP_005274280.1 27 3516
XM_005274224.1 XP_005274281.1 27 3513
XM_005274225.1 XP_005274282.1 26 3450
XM_005274226.1 XP_005274283.1 26 3447
XM_005274227.1 XP_005274284.1 26 3435
XM_005274228.1 XP_005274285.1 27 3378
XM_011545234.2 XP_011543536.1 27 3417
GenBank mRNA AB023171.2 - 27 3528
AF086762.1 - 26 3347
AK295013.1 - 15 1754

View Full Product Details