Gene Transcripts

Gene Symbol
CAPNS1
Entrez Gene ID
826
Gene Name
calpain small subunit 1
Gene Aliases
CALPAIN4, CANP, CANPS, CAPN4, CDPS, CSS1
Chromosome Location
Chr. 19: 36139926 - 36150353 on Build GRCh38
UniGene ID
Hs.515371
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003962.2 NP_001003962.1 5, 6 646
NM_001302632.1 NP_001289561.1 5, 6 553
NM_001302633.1 NP_001289562.1 4, 5 338
NM_001749.3 NP_001740.1 5, 6 649
XM_005259295.1 XP_005259352.1 5, 6 449
XM_005259296.1 XP_005259353.1 5, 6 452
XM_011527359.1 XP_011525661.1 4, 5 279
GenBank mRNA AK289606.1 - 5, 6 588
AY007141.1 - 5, 6 446
BC000592.2 - 5, 6 467 3347146
BC007779.2 - 5, 6 477 4301394
BC011903.2 - 5, 6 467 4120579
BC017308.2 - 5, 6 446 5089022
BC018931.2 - 5, 6 537 4303926
BC021933.1 - 5, 6 441 4872635
BC023643.2 - 5, 6 439 5017512
BC064998.1 - 5, 6 554 5785274
BQ891655.1 - 2, 3 110
BQ898664.1 - 5, 6 559
BT009775.1 - 4, 5 402
CN349928.1 - 4, 5 287
HY003903.1 - 4, 5 341
X04106.1 - 5, 6 560

View Full Product Details