Gene Transcripts

Gene Symbol
CHRM3
Entrez Gene ID
1131
Gene Name
cholinergic receptor muscarinic 3
Gene Aliases
EGBRS, HM3, PBS
Chromosome Location
Chr. 1: 239386565 - 239915450 on Build GRCh38
UniGene ID
Hs.155736
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000740.2 NP_000731.1 5 1034
XM_005273032.3 XP_005273089.1 6 1339
XM_011544041.2 XP_011542343.1 7 1438
XM_011544043.2 XP_011542345.1 9 1771
XM_011544044.2 XP_011542346.1 8 1542
XM_011544045.2 XP_011542347.1 8 1667
XM_011544046.2 XP_011542348.1 6 1375
XM_011544047.2 XP_011542349.1 6 1311
XM_017000152.1 XP_016855641.1 8 4364
XM_017000153.1 XP_016855642.1 7 1442
XM_017000154.1 XP_016855643.1 6 1136
XM_017000155.1 XP_016855644.1 7 1479
XM_017000156.1 XP_016855645.1 7 1443
XM_017000157.1 XP_016855646.1 5 1044
XM_017000158.1 XP_016855647.1 5 1276
XM_017000159.1 XP_016855648.1 5 1248
XM_017000160.1 XP_016855649.1 4 2098
XM_017000161.1 XP_016855650.1 6 1380
XM_017000162.1 XP_016855651.1 5 1212
XM_017000163.1 XP_016855652.1 4 5074
GenBank mRNA AF279779.1 - 5 1033
AF385589.1 - 1 255
AF498917.1 - 1 255
AK074575.1 - 1 412
BC096844.1 - 1 256
BC121026.2 - 2 398 40117429
BC121027.2 - 1 1588

View Full Product Details