Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004075.4 | NP_004066.1 | 9 | 2348 | |
XM_017018832.1 | XP_016874321.1 | 10 | 2483 | ||
GenBank mRNA | AK098615.1 | - | 4 | 752 | |
AK125915.1 | - | 11 | 2011 | ||
AK290552.1 | - | 9 | 1648 | ||
BC017466.1 | - | 9 | 1771 | ||
BC030519.1 | - | 9 | 2021 | 5169081 | |
D83702.1 | - | 9 | 2038 | ||
D84657.1 | - | 9 | 1452 |