Gene Transcripts
- Gene Symbol
- DOCK3
- Entrez Gene ID
- 1795
- Gene Name
- dedicator of cytokinesis 3
- Gene Aliases
- MOCA, PBP
- Chromosome Location
- Chr. 3: 50674969 - 51384198 on Build GRCh38
- UniGene ID
- Hs.476284
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004947.4 |
NP_004938.1
|
37, 38 |
3800 |
|
XM_005264914.3 |
XP_005264971.1
|
37, 38 |
4072 |
|
XM_005264915.3 |
XP_005264972.1
|
37, 38 |
4072 |
|
XM_005264916.3 |
XP_005264973.1
|
36, 37 |
3823 |
|
XM_005264917.3 |
XP_005264974.1
|
37, 38 |
4030 |
|
XM_005264918.3 |
XP_005264975.1
|
36, 37 |
3702 |
|
XM_006713008.3 |
XP_006713071.1
|
37, 38 |
4072 |
|
XM_006713009.3 |
XP_006713072.1
|
37, 38 |
4072 |
|
XM_006713010.3 |
XP_006713073.1
|
37, 38 |
3835 |
|
XM_011533441.2 |
XP_011531743.1
|
26, 27 |
4681 |
|
XM_011533443.2 |
XP_011531745.1
|
13, 14 |
2362 |
|
XM_011533444.2 |
XP_011531746.1
|
13, 14 |
2286 |
|
XM_011533445.2 |
XP_011531747.1
|
13, 14 |
2307 |
|
XM_017005825.1 |
XP_016861314.1
|
37, 38 |
3851 |
|
XM_017005826.1 |
XP_016861315.1
|
37, 38 |
3841 |
|
XM_017005827.1 |
XP_016861316.1
|
28, 29 |
2992 |
|
XM_017005828.1 |
XP_016861317.1
|
12, 13 |
1717 |
|
GenBank mRNA |
AB002297.1 |
- |
32, 33 |
3408 |
|
AY145303.1 |
- |
37, 38 |
3777 |
|
AY254099.1 |
- |
37, 38 |
3800 |
|
BC131746.1 |
- |
2, 3 |
168 |
|
View Full Product Details