Gene Transcripts

Gene Symbol
FANCC
Entrez Gene ID
2176
Gene Name
Fanconi anemia complementation group C
Gene Aliases
FA3, FAC, FACC
Chromosome Location
Chr. 9: 95099054 - 95317730 on Build GRCh38
UniGene ID
Hs.494529
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000136.2 NP_000127.2 8 1059
NM_001243743.1 NP_001230672.1 8 1001
NM_001243744.1 NP_001230673.1 8 1059
XM_005251802.3 XP_005251859.1 2 1161
XM_006717001.2 XP_006717064.1 7 876
XM_006717002.3 XP_006717065.1 8 1041
XM_006717004.3 XP_006717067.1 8 1041
XM_011518365.2 XP_011516667.1 8 1034
XM_011518366.2 XP_011516668.1 8 1041
XM_011518367.2 XP_011516669.1 9 1171
XM_017014452.1 XP_016869941.1 9 1186
XM_017014453.1 XP_016869942.1 9 1133
XM_017014454.1 XP_016869943.1 8 1060
GenBank mRNA AK222871.1 - 8 1021
AK312548.1 - 8 924
BC015748.1 - 8 1049 4864916
KU178051.1 - 7 797
KU178052.1 - 8 942
X66893.1 - 8 1052
X66894.1 - 8 970

View Full Product Details