Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000152.4 | NP_000143.2 | 13, 14 | 2287 | |
NM_001079803.2 | NP_001073271.1 | 14, 15 | 2102 | ||
NM_001079804.2 | NP_001073272.1 | 13, 14 | 2022 | ||
XM_005257193.2 | XP_005257250.1 | 14, 15 | 2088 | ||
XM_005257194.4 | XP_005257251.1 | 14, 15 | 2128 | ||
GenBank mRNA | AK299462.1 | - | 1939 | ||
BC040431.1 | - | 14, 15 | 2034 | 5739991 | |
DQ907243.1 | - | 12, 13 | 1890 | ||
M34424.1 | - | 12, 13 | 1922 | ||
Y00839.1 | - | 13, 14 | 2109 |