Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000847.4 | NP_000838.3 | 6 | 565 | |
XM_006715072.3 | XP_006715135.1 | 4 | 399 | ||
XM_011514533.2 | XP_011512835.1 | 3 | 391 | ||
XM_017010794.1 | XP_016866283.1 | 4 | 440 | ||
GenBank mRNA | AA846513.1 | - | 2 | 348 | |
AF020919.1 | - | 5 | 425 | ||
AF508266.1 | - | 5 | 486 | ||
AI199811.1 | - | 2 | 350 | ||
BC020619.1 | - | 6 | 552 | 4721622 | |
BG573805.1 | - | 462 |