Gene Transcripts
- Gene Symbol
- GYS1
- Entrez Gene ID
- 2997
- Gene Name
- glycogen synthase 1
- Gene Aliases
- GSY, GYS
- Chromosome Location
- Chr. 19: 48968125 - 48993353 on Build GRCh38
- UniGene ID
- Hs.386225
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161587.1 |
NP_001155059.1
|
13, 14 |
1852 |
|
NM_002103.4 |
NP_002094.2
|
14, 15 |
2044 |
|
NR_027763.1 |
|
13, 14 |
1862 |
|
GenBank mRNA |
AK223579.1 |
- |
14, 15 |
2000 |
|
AK291672.1 |
- |
14, 15 |
2000 |
|
AK294349.1 |
- |
9, 10 |
960 |
|
AK300369.1 |
- |
14, 15 |
1832 |
|
AK302738.1 |
- |
13, 14 |
1818 |
|
BC002617.2 |
- |
14, 15 |
1983 |
3143019 |
BC003182.1 |
- |
13, 14 |
1809 |
2958297 |
BC007688.2 |
- |
14, 15 |
1970 |
3953692 |
J04501.1 |
- |
14, 15 |
1963 |
|
U32573.1 |
- |
14, 15 |
1942 |
|
View Full Product Details