Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001527.3 | NP_001518.3 | 8, 9 | 1125 | |
NR_033441.1 | 9, 10 | 1153 | |||
NR_073443.1 | 8, 9 | 1031 | |||
XM_011535788.1 | XP_011534090.1 | 9, 10 | 877 | ||
XM_017010799.1 | XP_016866288.1 | 9, 10 | 1099 | ||
GenBank mRNA | AK092156.1 | - | 8, 9 | 1031 | |
AK097376.1 | - | 9, 10 | 1087 | ||
BC031055.1 | - | 8, 9 | 1089 | ||
U31814.1 | - | 8, 9 | 1042 |