Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019111.4 | NP_061984.2 | 3 | 497 | |
GenBank mRNA | AK301190.1 | - | 3 | 882 | |
BC032350.1 | - | 3 | 428 | 5214156 | |
BG548634.1 | - | 2 | 123 | ||
BM849755.1 | - | 3 | 498 | ||
M60334.1 | - | 3 | 415 |