Gene Transcripts
- Gene Symbol
- HLA-DRA
- Entrez Gene ID
- 3122
- Gene Name
- major histocompatibility complex, class II, DR alpha
- Gene Aliases
- HLA-DRA1, MLRW
- Chromosome Location
- Chr. 6: 32439842 - 32445046 on Build GRCh38
- UniGene ID
- Hs.520048
- Species
- Homo sapiens
- Gene Symbol
- HLA-DQA1
- Entrez Gene ID
- 3117
- Gene Name
- major histocompatibility complex, class II, DQ alpha 1
- Gene Aliases
- CD, CELIAC1, DQ-A1, GSE, HLA-DQA
- Chromosome Location
- Chr. 6: 32637403 - 32654846 on Build GRCh38
- UniGene ID
- Hs.387679
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF522250.1 |
- |
|
568 |
|
View Full Product Details