Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000423.2 | NP_000414.2 | 1 | 242 | |
GenBank mRNA | BC096294.4 | - | 1 | 231 | |
BC099643.1 | - | 1 | 231 | 40007309 | |
BC099644.3 | - | 1 | 231 | 40007310 | |
M99061.1 | - | 1 | 242 |