Gene Transcripts

Gene Symbol
MAP6
Entrez Gene ID
4135
Gene Name
microtubule associated protein 6
Gene Aliases
MAP6-N, MTAP6, N-STOP, STOP
Chromosome Location
Chr. 11: 75586914 - 75671108 on Build GRCh38
UniGene ID
Hs.585540
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033063.1 NP_149052.1 2 1003
NM_207577.1 NP_997460.1 2 1003
XM_006718556.3 XP_006718619.1 2 3642
XM_011545039.2 XP_011543341.1 2 3643
XM_017017755.1 XP_016873244.1 2 3542
XM_017017756.1 XP_016873245.1 2 105
XM_017017757.1 XP_016873246.1 3 172
XM_017017758.1 XP_016873247.1 2 139
GenBank mRNA AB058781.1 - 2 1003
AK123340.1 - 2 111
BC063860.1 - 2 669
BC139780.1 - 3 242 40122938
BC150254.1 - 2 1003 9007181

View Full Product Details